Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Formaldehyde 10% (v/v) in aqueous solution stabilized tissue fixative, for light and electron microscopy, buffered (Carson-Millonig Form. pH 7.4)

Supplier: Ricca Chemical
Danger
Synonyms: MethylaldehydeFormolFormalinParaformaldehyde aqueous solution

R3191000-20F R3191000-10F R3191000-1A R3191000-4A
RC31911EA 117.56 USD InStock
RC31911 RC31915 RC3191-32 RC319125
Formaldehyde 10% (v/v) in aqueous solution stabilized tissue fixative, for light and electron microscopy, buffered (Carson-Millonig Form. pH 7.4)
Formaldehyde


Formula: CH₂O
MW: 30.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
EINECS: 200-001-8
UN: 3334
ADR: 9,
Order Now
Specification Test Results
Appearance Clear, colorless, Formaldehyde odor
pH at 25 °C (traceable to NIST SRM 186 & 191) 7.40 ± 0.05 pH at 25.0 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty