Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Formalin-aceto-alcohol(F-A-A) mixture for rapid fixation of tissue

Supplier: Ricca Chemical
Danger

R3210000-500A R3210000-1A
RC321016EA 79.97 USD InStock
RC321016 RC3210-32
Formalin-aceto-alcohol(F-A-A) mixture for rapid fixation of tissue
Formalin-aceto-alcohol(F-A-A) mixture


Boiling Pt: ~ 82 °C
Density: 0.9200 g/cm³
Storage Temperature: Ambient
UN: 1987
ADR: 3,III
Order Now
Specification Test Results
Appearance Clear, colorless, Formaldehyde odor

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty