Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Carbol fuchsin (Ziehl-Neelsen)

Supplier: Ricca Chemical
Danger

R3220000-500C R3220000-120C
RC322016EA 130.45 USD InStock
RC322016 RC32204
Carbol fuchsin (Ziehl-Neelsen)
Carbol fuchsin
Fuchsin-Carbol, Ziehl-Neelsen Formulation, for Staining Acid Fast Organisms


Formula: C₂₁H₂₂ClN₃
MW: 351.88 g/mol
MDL Number: MFCD18071399
CAS Number: 4197-24-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty