Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Fuchsin solution decolourised R1

Supplier: Ricca Chemical

R3221000-1C R3221000-500C
RC3221-16EA 148.83 USD InStock
RC3221-16 RC3221-32
Fuchsin solution decolourised R1
Fuchsin solution decolorised R1


UN: 1789
ADR: 8,III
Order Now
Specification Test Results
Absorbance (traceable to NIST SRM 930) Conforms to control
Absorabnce at 545 nm Passes Test
Appearance Clear, colorless to slight yellow, odorless

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty