Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glycerin base USP Test Solution (TS)

Supplier: Ricca Chemical
Danger

R3287000-500A R3287000-120A
RC3287-4EA 73.57 USD InStock
RC3287-4 RC3287-16
Glycerin base USP Test Solution (TS)
Glycerin base


Order Now
Specification Test Results
Appearance Clear, colorless, odorless
Refractive Index Confirmed concentration

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty