Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glycerine 50% (v/v) in aqueous solution (1 + 1)

Supplier: Ricca Chemical
Warning
Synonyms: GlycerinGlyceritolGlycerolGlycyl alcoholGlyrolGlysaninOsmoglynPropanetriolTrihydroxypropane

R3290000-500A
RC329016EA 84.49 USD InStock
RC329016
Glycerine 50% (v/v) in aqueous solution (1 + 1)
Glycerine


Formula: HOCH₂CH(OH)CH₂OH
MW: 92.09 g/mol
Density: 1.1420 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00004722
CAS Number: 56-81-5
https://us-legacy.vwr.com/stibo/web/std.lang.all/47/74/734774.jpg
Order Now
Specification Test Results
Appearance Clear, colorless to slightly yellow, odorless.
Specific Gravity 1.140 ± 0.005 sp gr at 20.0 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty