Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid R1 250 g/L

Supplier: Ricca Chemical
Danger

R3557000-500A R3557000-1A
RC3557-16EA 48.69 USD InStock
RC3557-16 RC3557-32
Hydrochloric acid R1 250 g/L
Hydrochloric acid R1


Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.1500 g/cm³
Storage Temperature: Ambient
UN: 1789
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear, colorless, hydrochloric odor
Assay at 20 °C (traceable to NIST SRM 84) 250.0 ± 0.6 g/L HCl

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty