Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 1% (v/v) in aqueous solution (1 + 99)

Supplier: Ricca Chemical
Danger
Synonyms: Muriatic acid

R3560000-20F R3560000-500A R3560000-1A R3560000-4A
RC356032EA 48.1 USD InStock
RC356032 RC35605 RCR3560000-4A RC356016
Hydrochloric acid 1% (v/v) in aqueous solution (1 + 99)
Hydrochloric acid


Formula: HCl
MW: 36.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
ADR: 8,III
Order Now
Specification Test Results
Appearance Colorless liquid
Assay (vs. Sodium Hydroxide/Phenolphthalein) 0.96-1.04 % (v/v)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty