Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 50% (v/v) in aqueous solution (1 + 1)

Supplier: Ricca Chemical
Danger
Synonyms: Chlorohydric acidHydrochlorideHydrogen chloride gasMuriatic acid

R3580000-500A R3580000-20F R3580000-1A R3580000-4A R3580000-10F
RC3580-1EA 243.54 USD InStock
RC3580-1 RC358032 RC3580-2.5 RC358016 RCR3580000-20F
Hydrochloric acid 50% (v/v) in aqueous solution (1 + 1)
Hydrochloric acid


Formula: HCl
MW: 36.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II
Order Now
Specification Test Results
Appearance Colorless to slightly greenish-yellow liquid
Assay (vs. Sodium Hydroxide/Phenolphthalein) 49.6-50.4 % (v/v)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty