Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 50% (v/v) in aqueous solution

Supplier: Ricca Chemical
Synonyms: Muriatic acidHydrochloric acid solution

3580-5PT
RC3580-5PTEA 122.52 USD InStock
RC3580-5PT
Hydrochloric acid 50% (v/v) in aqueous solution
Hydrochloric acid


Formula: HCl
MW: 36.46 g/mol
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8,II
Order Now
Specification Test Results
Assay 50 ± 0.4 % (v/v)
NIST SRM 84

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty