Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 0.500 N (N/2) in 2-propanol

Supplier: Ricca Chemical
Danger

R3805000-4A R3805000-1A
RC38051EA 268.81 USD InStock
RC38051 RC380532
Hydrochloric acid 0.500 N (N/2) in 2-propanol
Hydrochloric acid


Formula: HCl
MW: 36.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1993
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear, colorless, HCl/IPA odor
Assay at 20 °C (traceable to NIST SRM 84) 0.5000 ± 0.0005 N at 20.0 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty