Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydroxylammonium chloride 30% (w/v) in aqueous solution

Supplier: Ricca Chemical
Synonyms: Hydroxylamine hydrochlorideHydroxylamine HCl

R3915000-120A R3915000-500A
RC3915-16EA 202.94 USD InStock
RC3915-16 RC3915-4
Hydroxylammonium chloride 30% (w/v) in aqueous solution
Hydroxylammonium chloride


Formula: NH₂OH·HCl
MW: 69.49 g/mol
MDL Number: MFCD00051089
CAS Number: 5470-11-1
UN: 2923
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear, colorless, odorless
Refractive Index Confirmed concentration

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty