Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hanus solution (Iodine bromine) for iodine absorption number of fats and oils

Supplier: Ricca Chemical
Danger
Synonyms: Iodine solution acc. to Hanus

R4100000-120C 4100-32 4100-16
RC4100-4EA 91.38 USD InStock
RC4100-4 RC410016 RC410032
Hanus solution (Iodine bromine) for iodine absorption number of fats and oils
Hanus solution (Iodine bromine)
Iodine-Bromine Solution, Hanus, for Iodine Absorption Number of Fats and Oils.

  • Meets AOAC standards


Formula: IBr
Density: 1.07
MDL Number: MFCD00011353
UN: 2920
ADR: 8 (3),II
Order Now
Specification Test Results
Appearance Dark brown liquid
Iodine/Bromine Ratio To Pass Test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty