Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Iodine D'Antoni solution, dark brown liquid

Supplier: Ricca Chemical

4103-16 4103-32 4103-4
RC4103-32EA 227.87 USD InStock
RC4103-32 RC4103-16 RC4103-4
Iodine D'Antoni solution, dark brown liquid
Iodine D'Antoni
D’Antoni’s Iodine stain is recommended for use in wet mount preparations for detection of protozoan cysts and helminth eggs and larvae.

Color: Dark brown liquid
REAGENTS (CLASSICAL FORMULA)*
Iodine (CAS 7553-56-2)...........................................................15.0 g
Potassium Iodide (CAS 7681-11-0).........................................10.0 g
Demineralized Water (CAS 7732-18-5) ...............................1000.0 ml
*Adjusted as required to meet performance standards.


UN: 3264
ADR: 8,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty