Synonyms:
ChloroiodideIodine chloride
4110-5PT
R4110000-500C
4110-32
4110-1
RC41101EA
718.94
USD
InStock
RC41101
RC411016
RC41105PT
RC411032
Iodine monochloride solution acc. Wijs USP Test Solution (TS) for iodine absorption number of fats and oils
Iodine monochloride
Meets AOAC standards for iodine absorption number.