Would you like to log in again?
Ionic strength adjustor (ISA) for pH indicating and adjusting, for use with ammonia selective electrode
Supplier: Ricca Chemical
R4128000-1A R4128000-500A
RC412832EA 128.41 USD InStock
RC412832 RC412816
Ionic strength adjustor (ISA) for pH indicating and adjusting, for use with ammonia selective electrode
Ionic strength adjustor (ISA)
Ionic strength adjustor, pH indicating and adjusting, for use with Ammonia Selective Electrode.
Order Now
Specification Test Results
Appearance Dark blue, methanol odor
ppm Ammonia No detectable Ammonia
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty