Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Kovac's indole reagent for detection of indole producing bacteria

Supplier: Ricca Chemical
Danger
Synonyms: 4-(Dimethylamino)benzaldehyde solution

R4260000-4C R4260000-500C R4260000-120C
RC426016EA 163.2 USD InStock
RC426016 RC4260-1 RC42604
Kovac's indole reagent for detection of indole producing bacteria
Kovac's indole reagent
Kovac's Aldehyde Reagent, for Detection of Indole Producing Bacteria


Formula: (CH₃)₂NC₆H₄CHO
MW: 149.19 g/mol
Boiling Pt: ~ 110 °C
Density: 0.9525 g/cm³
Flash Pt: ~ 40 °C
Storage Temperature: Ambient
MDL Number: MFCD00003381
CAS Number: 100-10-7
UN: 2920
ADR: 8 (3),II
Order Now
Specification Test Results
Appearance Clear, greenish-yellow, alcohol odor

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty