Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lead(II) acetate 9.5% (w/v) in aqueous solution USP Test Solution (TS)

Supplier: Ricca Chemical
Synonyms: Lead (II) acetate Acetic acid lead (II) salt

R4293000-500A R4293000-120A
RC4293-16EA 169.72 USD InStock
RC4293-16 RC4293-4
Lead(II) acetate 9.5% (w/v) in aqueous solution USP Test Solution (TS)
Lead (II) acetate


Formula: (H₃CCOO)₂Pb
MW: 325.29 g/mol
MDL Number: MFCD00012452
CAS Number: 301-04-2
UN: 1616
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 915) 9.50 ± 0.04 % (w/v)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty