Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lead(II) acetate solution 2% (w/v) alcoholic solution USP Test Solution (TS)

Supplier: Ricca Chemical
Synonyms: Acetic acid Lead(II) salt

R4293800-1A R4293800-500A
RC4293.8-16EA 154.48 USD InStock
RC4293.8-16 RC4293.8-32
Lead(II) acetate solution 2% (w/v) alcoholic solution USP Test Solution (TS)
Lead (II) acetate


MW: 325.29 g/mol
MDL Number: MFCD00012452
UN: 1616
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Clear, colorless, alcohol odor

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty