Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Magnesium chloride 51% (w/v) in aqueous solution

Supplier: Ricca Chemical

R4470000-500A R4470000-1A R4470000-4A
RC447016EA 144.23 USD InStock
RC447016 RCR4470000-4A RC447032
Magnesium chloride 51% (w/v) in aqueous solution
Magnesium chloride


Formula: MgCl₂
MW: 95.21 g/mol
Boiling Pt: ~ 100 °C
Density: 1.1791 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011106
CAS Number: 7786-30-3
Order Now
Specification Test Results
Appearance Colorless liquid
Assay (by Refractive Index) 50.5-51.5 % (w/v)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty