Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Magnesium sulfate solution 250 g/L in sulfuric acid 2N

Supplier: Ricca Chemical
Danger
Synonyms: Magnesium(II) sulfateBitter saltSulfuric acid magnesium salt (1:1)Epsom salt

R4550000-1A R4550000-500A
RC4550-16EA 71.45 USD InStock
RC4550-16 RC4550-32
Magnesium sulfate solution 250 g/L in sulfuric acid 2N
Magnesium sulphate


Formula: MgSO₄
MW: 120.37 g/mol
Boiling Pt: ~ 100 °C
Density: 1.1200 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011110
CAS Number: 7487-88-9
UN: 3264
ADR: 8,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/17/16640617.jpg
Order Now
Specification Test Results
Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 915) 250 ± 1 g/L

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty