Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mercury(II) chloride USP Test Solution (TS)

Supplier: Ricca Chemical
Synonyms: Mercury dichlorideMercuric chlorideMercury Chloride

R4660000-500A
RC4660-16EA 188.99 USD InStock
RC4660-16
Mercury(II) chloride USP Test Solution (TS)
Mercury (II) chloride


Formula: HgCl₂
MW: 271.5 g/mol
MDL Number: MFCD00011041
CAS Number: 7487-94-7
UN: 1624
ADR: 6.1,II
Order Now
Specification Test Results
Appearance Clear, colorless, odorless

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty