Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mercury(II) nitrate 0.282 N

Supplier: Ricca Chemical
Synonyms: Mercury dinitrateMercuric nitrate

R4750000-1C R4750000-500C R4750000-4C
RC475032EA 156.39 USD InStock
RC475032 RC47501 RC4750-16
Mercury(II) nitrate 0.282 N
Mercury (II) nitrate
1 ml = 10 mg Cl-


Formula: HgN₂O₆
MW: 324.60 g/mol
MDL Number: MFCD00011038
CAS Number: 10045-94-0
UN: 1625
ADR: 6.1,II
Order Now
Specification Test Results
Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 999) 0.2820 ± 0.0001 N at 20.0 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty