Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mercuric sulfate Denigès' reagent USP Test Solution (TS)

Supplier: Ricca Chemical
Danger

R4779000-120A R4779000-500A
RC4779-4EA 105.52 USD InStock
RC4779-4 RC4779-16
Mercuric sulfate Denigès' reagent USP Test Solution (TS)
Mercuric sulfate Denigès' reagent


Density: 1.2956 g/cm³
Storage Temperature: Ambient
UN: 2922
ADR: 8 (6.1),II
Order Now
Specification Test Results
Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 999) 0.491 ± 0.010 N at 20.0 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty