Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mercuric sulfate solution (red mercuric oxide) 80 g/L in 6 N Sulfuric Acid

Supplier: Ricca Chemical
Danger

R4780000-500A R4780000-1A
RC4780-16EA 369.4 USD InStock
RC4780-16 RC4780-32
Mercuric sulfate solution (red mercuric oxide) 80 g/L in 6 N Sulfuric Acid
Mercuric sulfate solution (red mercuric oxide)


Density: 1.2000 g/cm³
Storage Temperature: Ambient
UN: 2922
ADR: 8 (6.1),II
Order Now
Specification Test Results
Appearance Clear, colorless, odorless

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty