Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Methylamino)phenol sulfate USP reference solution (RS)

Supplier: Ricca Chemical
Synonyms: 4-(Methylamino)phenol hemisulfate saltMetol4-(Methylamino)phenol hemisulphatep-(Methylamino)phenol sulphate4-(Methylamino)phenol hemisulfate4-Methylaminophenolsulphatep-(Methylamino)phenol sulfate4-(Methylamino)phenol hemisulphate salt4-Methylaminophenolsulfate4-Methylaminophenol sulfate4-Methylaminophenol sulphate

R4839000-120C R4839000-500C
RC4839-16EA 82.75 USD InStock
RC4839-16 RC4839-4
4-(Methylamino)phenol sulfate USP reference solution (RS)
4-(Methylamino)phenol sulfate


Formula: C₁₄H₂₀N₂O₆S
MW: 344.39 g/mol
MDL Number: MFCD00013140
CAS Number: 55-55-0
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Clear, pale green, Bisulfite odor
Suitability for Use Blue in the presence of Phosphate

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty