Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methylene blue 1% (w/v) in aqueous solution

Supplier: Ricca Chemical

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: Methylthioninium chloride 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride

R4850000-500A R4850000-120A R4850000-1A
RC4850-32EA 108.54 USD InStock
RC4850-32 RC485016 RC48504
Methylene blue 1% (w/v) in aqueous solution
Methylene blue


Formula: C₁₆H₁₈ClN₃S
MW: 319.86 g/mol
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0050 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00150008
CAS Number: 61-73-4
Order Now
Specification Test Results
Absorbance (traceable to NIST SRM 930) Conforms to control
Appearance Dark blue, odorless

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty