Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methylene blue solution 0.05% (w/v) alcoholic solution USP Test Solution (TS)

Supplier: Ricca Chemical

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: Basic Blue 9Swiss BlueMethylthioninium chlorideMethylene blue chloride3,7-Bis(dimethylamino)phenothiazin-5-ium chloride

R4862000-120A R4862000-500A
RC4862-4EA 68.87 USD InStock
RC4862-4 RC4862-16
Methylene blue solution 0.05% (w/v) alcoholic solution USP Test Solution (TS)
Methylene blue


Formula: C₁₆H₁₈ClN₃S
MW: 319.86 g/mol
Boiling Pt: ~ 77 °C
Density: 0.8000 g/cm³
Flash Pt: ~ 15.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00150008
CAS Number: 61-73-4
UN: 1987
ADR: 3,II
Order Now
Specification Test Results
Absorbance (traceable to NIST SRM 930) Absorbance Maxima
Appearance Clear, blue, Ethanol odor

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty