Synonyms:
Basic Blue 9
, Swiss Blue
, Methylthioninium chloride
, Methylene blue chloride
, 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride
R4880000-500A
R4880000-120A
RC48804EA
50.7
USD
InStock
RC48804
RC488016
Methylene blue (Loeffler) for staining and differentiation of acid fast organisms in smears
Methylene blue
Methylene blue, Loeffler formulation