Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methylene blue solution II 0.05 mL (1 drop) = 0.1 mg/L Sulfide

Supplier: Ricca Chemical
Synonyms: Swiss BlueBasic blue 9Methylthioninium chlorideCI 520153,7-Bis(dimethylamino)phenothiazin-5-ium chloride

R4910000-500A R4910000-120A
RC491016EA 183.36 USD InStock
RC491016 RC4910-4
Methylene blue solution II 0.05 mL (1 drop) = 0.1 mg/L Sulfide
Methylene blue trihydrate


Formula: C₁₆H₁₈ClN₃S·3H₂O
MW: 373.9 g/mol
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0000 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00150008
CAS Number: 7220-79-3
Order Now
Specification Test Results
Appearance Clear, dark blue, odorless
Certified Concentration 0.05 mL = 0.1 mg/L Sulfide

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty