Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl orange 0.05% (w/v) in aqueous solution indicator

Supplier: Ricca Chemical
Synonyms: 4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulphonic acid, sodium saltMethyl orange sodium saltHelianthine4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulfonic acid, sodium salt

R4980000-120A R4980000-500A
RC498016EA 51.65 USD InStock
RC498016 RC49804
Methyl orange 0.05% (w/v) in aqueous solution indicator
Methyl orange


Formula: C₁₄H₁₄N₃NaO₃S
MW: 327.34 g/mol
MDL Number: MFCD00007502
CAS Number: 547-58-0
UN: 3143
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Clear, orange, odorless
Suitability for Use Yellow (pH 4.4) - Pinkish-Orange (pH 3.2) - Pink (pH 3.2)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty