Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Alizarin red S (sodium salt) 1% (w/v) in aqueous solution indicator

Supplier: Ricca Chemical
Warning
Synonyms: Sodium 3,4-dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-sulfonateSodium 3,4-dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-sulphonateAlizarin sulfonic acid disodium saltC.I. 58005Alizarin sulphonic acid disodium salt

R0500000-120A R0500000-500A
RC5004EA 35.72 USD InStock
RC5004 RC50016
Alizarin red S (sodium salt) 1% (w/v) in aqueous solution indicator
Alizarin Red S (sodium salt)


Formula: C₁₄H₇NaO₇S
MW: 342.26 g/mol
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0000 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00013049
CAS Number: 130-22-3
Order Now
Specification Test Results
Appearance Dark, reddish-orange color, odorless
Suitability for Use Yellow (pH 4.6) - Pink (pH 6.0)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty