Would you like to log in again?
Acetate Buffer solutions, pH 4.00, Ricca Chemical Company
Supplier: RICCA Chemical Company
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
R0050000-1A R0050000-20F R0050000-4A
RC501EA 281.87 USD InStock
RC501 RC505 RC5032
Acetate Buffer solutions, pH 4.00, Ricca Chemical Company
Buffers pH Calibration Buffers
Acetate buffer, pH 4.0, for residual chlorine analysis
Standardized at 25°C against chemicals whose certification is traceable to NIST.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty