Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl purple solution indicator

Supplier: Ricca Chemical
Synonyms: Fleisher's Indicator Solution

R5040000-500C R5040000-1C R5040000-120C
RC504016EA 54.09 USD InStock
RC504016 RC504032 RC50404
Methyl purple solution indicator
Methyl purple
Aqueous.


Formula: C₆₇H₆₂N₈Na₄O₁₄S₃
MW: 1391.41 g/mol
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0050 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00070843
CAS Number: 1340-02-9
Order Now
Specification Test Results
Appearance Clear, dark, medium green, odorless
Suitability for Use Purple (pH 4.8) - Green (pH 5.4), with Gray warning color

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty