Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl violet 2B 0.02% (w/v) in aqueous solution indicator

Supplier: Ricca Chemical
Synonyms: Basic Violet 14-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]methylammonium chloride

R5100000-500A R5100000-120A
RC51004EA 52.36 USD InStock
RC51004 RC510016
Methyl violet 2B 0.02% (w/v) in aqueous solution indicator
Methyl Violet 2B


Formula: C₂₄H₂₈ClN₃
MW: 393.96 g/mol
MDL Number: MFCD00012568
CAS Number: 8004-87-3
UN: 3077
ADR: 9,III
Order Now
Specification Test Results
Appearance Clear, purple, odorless
Suitability for Use Yellow (pH 3.2)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty