Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl red-methylene blue solution mixed indicator

Supplier: Ricca Chemical
Danger

R5164000-120A R5164000-500A
RC5164-16EA 83.92 USD InStock
RC5164-16 RC5164-4
Methyl red-methylene blue solution mixed indicator
Methyl red-methylene blue


Boiling Pt: ~ 77 °C to ~ 77 °C
Density: 0.8000 g/cm³
Storage Temperature: Ambient
UN: 1987
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear, dark green, alcohol odor
Suitability for Use Purple (pH 6.2)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty