Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Murexide 0.2% (w/w) in sodium chloride indicator

Supplier: Ricca Chemical
Synonyms: Ammonium 5-(2,4,6-trioxoperhydropyrimidin-5-ylideneamino)barbiturate Ammonium purpurate Purpuric acid ammonium salt

R5220000-500A5
RC5220-500EA 422.38 USD InStock
RC5220-500
Murexide 0.2% (w/w) in sodium chloride indicator
Murexide


Formula: C₈H₈N₆O₆
MW: 284.19 g/mol
Boiling Pt: 1461 °C
Melting Pt: 801 °C
Density: 2.1000 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00012777
CAS Number: 3051-09-0
Order Now
Specification Test Results
Appearance Rosy pink powder
Suitability for Use Pink (Excess Calcium) - Purple (Excess EDTA)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty