Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Naphtholbenzeine 1% (w/v) in titration solvent indicator

Supplier: Ricca Chemical
Danger
Synonyms: alpha-Naphtholbenzeinα-Naphtholbenzein4-(α-(4-Hydroxy-1-naphthyl)benzylidene)naphthalen-1(4H)-one4-Hydroxy-α-(4-hydroxynaphthyl)-α-phenylnaphthalene-1-methanolα-Naphtholbenzeinep-Naphtholbenzeine1-NaphtholbenzeinBis(4-hydroxy-1-naphthyl)phenylmethanolp-Naphtholbenzein

R5225000-1C R5225000-120C
RC5225-4EA 86.89 USD InStock
RC5225-4 RC5225-32
1-Naphtholbenzeine 1% (w/v) in titration solvent indicator
1-Naphtholbenzeine


Formula: C₂₇H₁₈O₂
MW: 374.44 g/mol
Boiling Pt: ~ 90 °C
Density: 0.8200 g/cm³
Flash Pt: < 5 °C
Storage Temperature: Ambient
MDL Number: MFCD00078492
CAS Number: 145-50-6
UN: 1993
ADR: 3,II
Order Now
Specification Test Results
Appearance Dark red, Toluene odor
Suitability for Use Yellow - Orange to Green/Green-Brown (Acidic) - Green/Brown to Orange (Basic)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty