Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Neocuproine 0.1% (w/v) in methanol, Laboratory Reagent

Supplier: Ricca Chemical
Danger
Synonyms: 2,9-Dimethyl-1,10-phenanthroline

R5240000-500A R5240000-120A
RC524016EA 149.69 USD InStock
RC524016 RC5240-4
Neocuproine 0.1% (w/v) in methanol, Laboratory Reagent
Neocuproine


Formula: C₁₄H₁₂N₂
MW: 208.26 g/mol
Boiling Pt: ~ 64.5 °C
Melting Pt: –98 °C
Density: 0.8000 g/cm³
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00004973
CAS Number: 484-11-7
UN: 1993
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear, colorless, Methanol odor

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty