Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nessler reagent USP XXV

Supplier: Ricca Chemical
Danger
Synonyms: Nota 1Potassium tetraiodomercurateMercury (II) potassium iodidePotassium tetraiodomercurate – Potassium hydroxide solutionPotassium mercuric Iodide

R5250000-500B R5255000-1B R5250000-1B
RC525016EA 203.6 USD InStock
RC525016 RC5255-32 RC525032
Nessler reagent USP XXV
Nessler's reagent
(Mercuric - Potassium Iodide TS, Alkaline), USP 25 Formulation


MW: 786.4 g/mol
Boiling Pt: ∼100 °C (1013 hPa)
Density: 1.2 g/cm³ (20 °C)
MDL Number: MFCD00049656
UN: 2922
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear, light yellow, odorless
Suitability for Use Passes Test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty