Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nessler reagent Folin-Wu Modified for determination of non-protein nitrogen (NPN)

Supplier: Ricca Chemical
Danger
Synonyms: Nota 1Potassium tetraiodomercurateMercury (II) potassium iodidePotassium tetraiodomercurate – Potassium hydroxide solutionPotassium mercuric Iodide

R5260000-500B
RC5260-16EA 157.46 USD InStock
RC5260-16
Nessler reagent Folin-Wu Modified for determination of non-protein nitrogen (NPN)
Nessler's reagent
Folin-Wu modified for determination of non-protein nitrogen.


MW: 786.4 g/mol
Boiling Pt: 356.6 °C (1013 hPa)
Melting Pt: –38.9 °C
Density: 13.6 g/cm³ (20 °C)
MDL Number: MFCD00049656
EINECS: 231-990-4
UN: 2922
ADR: 8,II
Merck Index: 12,07862
Order Now
Specification Test Results
Ammonia Sensitivity 0.1 mg/L Ammonia Nitrogen
Appearance Clear, light yellow, odorless

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty