Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Neutral red 0.1% (w/v) in 50% alcohol USP Test Solution (TS)

Supplier: Ricca Chemical
Danger
Synonyms: N8,N8,3-Trimethyl-2,8-phenazinediamine monohydrochloride Basic Red 5

R5271000-120A R5271000-500A
RC5271-4EA 44.84 USD InStock
RC5271-4 RC5271-16
Neutral red 0.1% (w/v) in 50% alcohol USP Test Solution (TS)
Neutral red


Formula: C₁₅H₁₇ClN₄
MW: 288.78 g/mol
Boiling Pt: ~ 82 °C
Density: 0.9000 g/cm³
Flash Pt: 24 °C
Storage Temperature: Ambient
MDL Number: MFCD00012651
CAS Number: 553-24-2
UN: 1987
ADR: 3,III
Order Now
Specification Test Results
Appearance Clear, red, alcohol odor
Suitability for Use Red (pH 6.8) - Orange (pH 8.0)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty