Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

New methylene Blue N for reticulocyte count

Supplier: Ricca Chemical
Synonyms: Basic Blue 24Methylene Blue N

R5285000-500A R5285000-120A
RC528516EA 191.64 USD InStock
RC528516 RC52854
New methylene Blue N for reticulocyte count
New methylene Blue N
For reticulocytes.


Formula: C₁₈H₂₂ClN₃S
MW: 347.91 g/mol
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0000 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00135878
CAS Number: 1934-16-3
Order Now
Specification Test Results
Absorbance (traceable to NIST SRM 930) Conforms to control
Appearance Deep, purple odorless

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty