Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nitric acid, dilute R 125 g/L

Supplier: Ricca Chemical
Danger

R5330000-500C R5330000-1C
RC5330-32EA 158.94 USD InStock
RC5330-32 RC5330-16
Nitric acid, dilute R 125 g/L
Nitric acid, dilute R


Formula: HNO3
MW: 63,012 g/mol
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0660 g/cm³
Storage Temperature: Ambient
UN: 3264
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear, colorless, Nitric odor
Assay at 20 °C (traceable to NIST SRM 84) 125.0 ± 0.1 g/L HNO3

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty