Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Alkaline iodide azide for determination of dissolved oxygen in water

Supplier: Ricca Chemical
Danger

541-16 541-32
RC54116EA 466.75 USD InStock
RC54116 RC54132
Alkaline iodide azide for determination of dissolved oxygen in water
Alkaline iodide azide


Boiling Pt: ~ 104 °C
Density: 1.7500 g/cm³
Storage Temperature: Ambient
UN: 1824
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear to turbid, odorless
Free Iodine none detectable

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty