Would you like to log in again?
Phenanthroline ferrous sulfate (orthophenanthroline TS) 0.025 M
Supplier: Ricca Chemical
R5464800-120C R5464800-500C
RC5464.8-4EA 99.61 USD InStock
RC5464.8-4 RC5464.8-16
Phenanthroline ferrous sulfate (orthophenanthroline TS) 0.025 M
Phenanthroline ferrous sulfate (orthophenanthroline TS)
Mixture 1,10-Phenanthroline Monohydrate, Ferrous Sulfate Heptahydrate in aqueous solution.
Order Now
Specification Test Results
Appearance brownish orange to blood red, odorless
Suitability for Use Green to Bluish Green (Reduced) - Redish (Oxidized)
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty