Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Oxalic acid 5% (w/v) in aqueous solution analytical reagent

Supplier: Ricca Chemical
Danger

R5465500-500A R5465500-1A
RC5465.5-32EA 70.25 USD InStock
RC5465.5-32 RC5465.5-16
Oxalic acid 5% (w/v) in aqueous solution analytical reagent
Oxalic acid


Formula: C₂H₂O₄
MW: 90.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002573
CAS Number: 144-62-7
EINECS: 205-634-3
UN: 3265
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear, colorless, odorless
Refractive Index 5.0 ± 0.5 % (w/v)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty