Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Oxalic acid 7.5% (w/v) in aqueous solution

Supplier: Ricca Chemical

R5467000-500A R5467000-1A R5467000-4A
RC5467-32EA 98.46 USD InStock
RC5467-32 RC5467-1 RC546716
Oxalic acid 7.5% (w/v) in aqueous solution
Oxalic acid


Formula: C₂H₂O₄
MW: 90.04 g/mol
MDL Number: MFCD00002573
CAS Number: 144-62-7
UN: 3265
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear, colorless, odorless
Refractive Index 7.5 ± 0.6 % (w/v)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty