Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Papanicolaou's solution EA 65 stain for cytology

Supplier: Ricca Chemical
Danger
Synonyms: EA 36

R5505000-500A R5505000-1A R5505000-4A
RC5505-32EA 166.3 USD InStock
RC5505-32 RC55051 RC550516
Papanicolaou's solution EA 65 stain for cytology
Papanicolaou's solution EA 65
Papanicolaou Stain, EA-65 Formula, Counterstain for use with Hematoxylin Stains in the Papanicolaou Staining Procedure


Boiling Pt: ~ 80 °C
Density: 0.8053 g/cm³
Flash Pt: ~ 16 °C
Storage Temperature: Ambient
UN: 1987
ADR: 3,II
Order Now
Specification Test Results
Absorbance (traceable to NIST SRM 930) Conforms to control
Appearance Murky green, alcohol odor

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty