Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Perchloric acid 0.100 N (N/10) in glacial acetic acid for non-aqueous titration

Supplier: Ricca Chemical

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger

R5515000-4C R5515000-1C R5515000-500C R5515000-20F R5515000-2.5D
RCR55150002.5DEA 265.43 USD InStock
RCR55150002.5D RCR5515000-20F RCR5515000-1C RC55151 RCR5515000500C
Perchloric acid 0.100 N (N/10) in glacial acetic acid for non-aqueous titration
Perchloric acid


Formula: HClO₄
MW: 100.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011325
CAS Number: 7601-90-3
UN: 2789
ADR: 8 (3),II
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/22/16640622.jpg
Order Now
Specification Test Results
Appearance Colorless to light yellow liquid
Assay (vs. Potassium Hydrogen Phthalate/Crystal Violet) 0.0999-0.1001 N at 20°C
Water (H₂O) 0.020-0.050 % (w/w)

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty